Novus Biologicals
Manufacturer Code:NBP2266660.1ML
Catalog # NB226601M
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 12A5 |
Immunogen | CD3 mAb-stimulated CD4/CD25 T cells |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: Folate receptor 4; folate receptor 4 (delta) homolog; folate receptor 4 (delta) homolog (mouse) Folbp3 probable folate receptor delta; folate receptor 4, delta (putative); Folate receptor delta; FR-delta; IZUMO1 receptor protein JUNO; probable folate receptor delta; Sperm-egg fusion protein Juno
Gene Aliases: Folbp3; FOLR4; IZUMO1R; JUNO
UniProt ID: (Human) A6ND01
Entrez Gene ID: (Human) 390243
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.