Novus Biologicals
Manufacturer Code:NBP270763JF549
Catalog # N270763J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G6 |
Immunogen | Recombinant protein fragment corresponding to amino acids 20-221 of human FOLR2 (NP_000794) produced in E.coli. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BETA-HFR FBP FBP/PL-1 Folate receptor 2 folate receptor 2 (fetal) folate receptor beta folate receptor beta Folate receptor fetal/placental folate-binding protein fetal/placental FR-BETA FR-P3 Placental folate-binding protein; FBP; Folate receptor 2; folate receptor 2 (fetal); Folate receptor beta; folate receptor, beta; Folate receptor, fetal/placental; folate-binding protein, fetal/placental; FR-beta; Placental folate-binding protein
Gene Aliases: BETA-HFR; FBP; FBP/PL-1; FOLR2; FR-BETA; FR-P3
UniProt ID: (Human) P14207
Entrez Gene ID: (Human) 2350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.