Invitrogen
Manufacturer Code:PA545768
Catalog # PIPA545768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FOLR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FBP; folate binding protein; folate binding protein 2; Folate receptor 2; folate receptor 2 (fetal); folate receptor 2 (fetal) BETA-HFR FBP/PL-1 FR-BETA FR-P3; Folate receptor beta; folate receptor, beta; Folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; FR-beta; Placental folate-binding protein
Gene Aliases: BETA-HFR; FBP; FBP/PL-1; FBP2; Folbp-2; Folbp2; FOLR2; FR-BETA; FR-P3
UniProt ID: (Human) P14207, (Mouse) Q05685
Entrez Gene ID: (Pig) 396853, (Rabbit) 100341917, (Bovine) 507672, (Dog) 476816, (Horse) 100066065, (Human) 2350, (Rat) 293154, (Mouse) 14276
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.