R&D Systems
Manufacturer Code:AF5697
Catalog # AF5697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human FOLR2 Gln22-His228 Accession # P14207 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BETA-HFR FBP FBP/PL-1 Folate receptor 2 folate receptor 2 (fetal) folate receptor beta folate receptor beta Folate receptor fetal/placental folate-binding protein fetal/placental FR-BETA FR-P3 Placental folate-binding protein; FBP; Folate receptor 2; folate receptor 2 (fetal); Folate receptor beta; folate receptor, beta; Folate receptor, fetal/placental; folate-binding protein, fetal/placental; FR-beta; Placental folate-binding protein
Gene Aliases: BETA-HFR; FBP; FBP/PL-1; FOLR2; FR-BETA; FR-P3
UniProt ID: (Human) P14207
Entrez Gene ID: (Human) 2350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.