Novus Biologicals
Manufacturer Code:NB10062577V
Catalog # NB10062577V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SF55 (BGN/1210/55) |
Immunogen | Folate binding protein from bovine milk |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adult folate-binding protein; Adult folate-binding protein FBP folate binding protein Folate receptor 1 folate receptor 1 (adult) folate receptor alpha Folate receptor adult FOLR FR-alpha KB cells FBP Ovarian tumor-associated antigen MOv18; FBP; folate binding protein; Folate receptor 1; folate receptor 1 (adult); Folate receptor alpha; Folate receptor, adult; FR-alpha; KB cells FBP; Ovarian tumor-associated antigen MOv18
Gene Aliases: FBP; FOLR; FOLR1
UniProt ID: (Human) P15328
Entrez Gene ID: (Human) 2348
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.