Proteintech
Manufacturer Code:603071IG150UL
Catalog # 50-173-6400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B4B7 |
Immunogen | FOLR1 Fusion Protein Ag19959 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adult folate binding protein FBP Folate receptor 1 folate receptor 1 (adult) Folate receptor alpha Folate receptor adult FOLR FOLR1 FR alpha KB cells FBP MOv18; Adult folate-binding protein; FBP; folate binding protein; Folate receptor 1; folate receptor 1 (adult); Folate receptor alpha; Folate receptor, adult; FR-alpha; KB cells FBP; Ovarian tumor-associated antigen MOv18
Gene Aliases: FBP; FOLR; FOLR1
UniProt ID: (Human) P15328
Entrez Gene ID: (Human) 2348
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.