Novus Biologicals
Manufacturer Code:NBP132895
Catalog # NBP132895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human FOG2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FOG-2; friend of GATA 2; Friend of GATA protein 2; Friend of GATA2; transcription factor GATA4 modulator of zinc finger protein 89B zinc finger protein multitype 2; transcription factor GATA4, modulator of; Zinc finger protein 89B; Zinc finger protein multitype 2; Zinc finger protein ZFPM2; zinc finger protein, multitype 2
Gene Aliases: DIH3; FOG2; hFOG-2; SRXY9; ZC2HC11B; ZFPM2; ZNF89B
UniProt ID: (Human) Q8WW38
Entrez Gene ID: (Human) 23414
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.