Invitrogen
Manufacturer Code:PA512159
Catalog # PIPA512159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 6-36 amino acids from the N-terminal region of human FNTB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAAX farnesyltransferase subunit beta; FTase-beta; Protein farnesyltransferase subunit beta; Protein farnesyltransferase subunit beta FTase-beta EC 2.5.1.58 CAAX farnesyltransferase subunit beta RAS proteins prenyltransferase beta FNTB; Ras proteins prenyltransferase subunit beta
Gene Aliases: FNTB; FPTB
UniProt ID: (Human) P49356
Entrez Gene ID: (Human) 2342
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.