Invitrogen
Manufacturer Code:PA512169
Catalog # PIPA512169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 88-118 amino acids from the central region of human FNTA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAAX farnesyltransferase subunit alpha; farnesyl-protein transferase alpha-subunit; FTase-alpha; GGTase-I-alpha; Protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha; Protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha CAAX farnesyltransferase subunit alpha Ras proteins prenyltransferase alpha FTase-alpha Type I protein geranyl-geranyltransferase subunit alpha GGTase-I-alpha; protein prenyltransferase alpha subunit repeat containing 2; Ras proteins prenyltransferase subunit alpha; type I protein geranyl-geranyltransferase alpha subunit; Type I protein geranyl-geranyltransferase subunit alpha
Gene Aliases: FNTA; FPTA; FTA; PGGT1A; PTAR2
UniProt ID: (Human) P49354, (Mouse) Q61239
Entrez Gene ID: (Human) 2339, (Mouse) 14272
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.