Invitrogen
Manufacturer Code:PA552474
Catalog # PIPA552474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FNDC3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Factor for adipocyte differentiation 104; fibronectin domain-containing protein 3b; Fibronectin type III domain-containing protein 3B; HCV NS5A-binding protein 37
Gene Aliases: 1600019O04Rik; AW550168; FAD104; FNDC3B; Kiaa4164; mKIAA4164; NS5ABP37; PRO4979; RGD1311673; UNQ2421/PRO4979/PRO34274; YVTM2421
UniProt ID: (Human) Q53EP0, (Mouse) Q6NWW9
Entrez Gene ID: (Human) 64778, (Rat) 294925, (Mouse) 72007
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.