Invitrogen
Manufacturer Code:PA572247
Catalog # PIPA572247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 24-58 amino acids from human FN3KRP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FN3K-related protein; FN3K-RP; fructosamine 3-kinase-related protein; Fructosamine-3-kinase-related protein; Ketosamine-3-kinase; Ketosamine-3-kinase 271- Fructosamine-3-kinase-related protein FN3K-RP FN3K-related protein FN3KRP; ortholog of human fructosamine-3-kinase-related protein FN3KRP; Protein-psicosamine 3-kinase FN3KRP; testis secretory sperm-binding protein Li 211a
Gene Aliases: FN3K-RP; FN3KL; FN3KRP
UniProt ID: (Human) Q9HA64, (Mouse) Q8K274
Entrez Gene ID: (Human) 79672, (Mouse) 238024
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.