Invitrogen
Manufacturer Code:PA530819
Catalog # PIPA530819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 64 and 279 of Human FN3KRP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FN3K-related protein; FN3K-RP; FN3KL FN3K-RP FN3K-related protein fructosamine-3-kinase-related protein ketosamine-3-kinase; Fructosamine-3-kinase-related protein; Ketosamine-3-kinase; Protein-psicosamine 3-kinase FN3KRP; testis secretory sperm-binding protein Li 211a
Gene Aliases: FN3KL; FN3KRP
UniProt ID: (Human) Q9HA64
Entrez Gene ID: (Human) 79672
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.