Invitrogen
Manufacturer Code:PA5109734
Catalog # PIPA5109734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-309 of human FN3KRP (NP_078895.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FN3K-related protein; FN3K-RP; FN3KL Fn3krp FN3K-RP Fructosamine-3-kinase-related protein Ketosamine-3-kinase Protein-psicosamine 3-kinase FN3KRP RGD1304570; fructosamine 3-kinase-related protein; Fructosamine-3-kinase-related protein; Ketosamine-3-kinase; ortholog of human fructosamine-3-kinase-related protein FN3KRP; Protein-psicosamine 3-kinase FN3KRP; testis secretory sperm-binding protein Li 211a
Gene Aliases: FN3K-RP; FN3KL; FN3KRP; RGD1304570
UniProt ID: (Human) Q9HA64, (Mouse) Q8K274
Entrez Gene ID: (Human) 79672, (Rat) 303755, (Mouse) 238024
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.