Proteintech
Manufacturer Code:163121AP150UL
Catalog # 50-173-0868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FN3KRP Fusion Protein Ag9347 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FN3K related protein FN3K RP FN3KL FN3KRP Ketosamine 3 kinase; FN3K-related protein; FN3K-RP; fructosamine 3-kinase-related protein; Fructosamine-3-kinase-related protein; Ketosamine-3-kinase; ortholog of human fructosamine-3-kinase-related protein FN3KRP; Protein-psicosamine 3-kinase FN3KRP; testis secretory sperm-binding protein Li 211a
Gene Aliases: FN3K-RP; FN3KL; FN3KRP; RGD1304570
UniProt ID: (Human) Q9HA64, (Mouse) Q8K274
Entrez Gene ID: (Human) 79672, (Rat) 303755, (Mouse) 238024
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.