Abnova
Manufacturer Code:H00079672K
Catalog # 89-200-554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human FN3KRP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human FN3KRP peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ12171; FN3K-related protein; FN3K-RP; Fructosamine-3-kinase-related protein; Ketosamine-3-kinase; Protein-psicosamine 3-kinase FN3KRP; testis secretory sperm-binding protein Li 211a
Gene Aliases: FN3KL; FN3KRP
UniProt ID: (Human) Q9HA64
Entrez Gene ID: (Human) 79672
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.