Invitrogen
Manufacturer Code:MA537616
Catalog # PIMA537616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 788CT24.5.1 |
Immunogen | purified His-tagged FN3K protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310074G21Rik FN3K Fnsk fructosamine 3 kinase fructosamine 3-kinase fructosamine-3-kinase Protein-psicosamine 3-kinase FN3K Protein-ribulosamine 3-kinase FN3K; Fructosamine-3-kinase; Protein-psicosamine 3-kinase FN3K; Protein-ribulosamine 3-kinase FN3K
Gene Aliases: FN3K
UniProt ID: (Human) Q9H479
Entrez Gene ID: (Human) 64122
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.