Invitrogen
Manufacturer Code:PA5114735
Catalog # PIPA5114735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FN3K corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310074G21Rik FN3K Fnsk fructosamine 3 kinase fructosamine 3-kinase fructosamine-3-kinase Protein-psicosamine 3-kinase FN3K Protein-ribulosamine 3-kinase FN3K; fructosamine 3-kinase; Fructosamine-3-kinase; Protein-psicosamine 3-kinase FN3K; Protein-ribulosamine 3-kinase FN3K
Gene Aliases: 2310074G21Rik; FN3K; Fnsk
UniProt ID: (Human) Q9H479, (Mouse) Q9ER35
Entrez Gene ID: (Human) 64122, (Rat) 498034, (Mouse) 63828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.