Abnova
Manufacturer Code:MAB21193
Catalog # 89-337-686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TV-1 |
Immunogen | T-cell lymphoma biopsy |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Mouse Monoclonal Antibody.
Protein Aliases: Anastellin; CIG; CIG DKFZp686F10164 DKFZp686H0342 DKFZp686I1370 DKFZp686O13149 ED-B FINC FN FNZ GFND GFND2 LETS MSF; Cold-insoluble globulin; Fibronectin; FN; migration-stimulating factor; Ugl-Y1; Ugl-Y2; Ugl-Y3
Gene Aliases: CIG; ED-B; FINC; FN; FN1; FNZ; GFND; GFND2; LETS; MSF
UniProt ID: (Human) P02751
Entrez Gene ID: (Human) 2335
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.