Abnova
Manufacturer Code:PAB27963
Catalog # 89-129-478
Quantity:
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A Synthetic peptide corresponding to amino acids of FN1 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage,aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. |
Chicken polyclonal antibody against synthetic peptide of FN1
For Research Use Only
Protein Aliases: Anastellin; CIG; Cold-insoluble globulin; DKFZp686F10164; DKFZp686H0342; DKFZp686I1370; DKFZp686O13149; ED-B; Fibronectin; FINC; FN; FNZ; GFND; GFND2; LETS; migration-stimulating factor; MSF; Ugl-Y1; Ugl-Y2; Ugl-Y3
Gene Aliases: CIG; ED-B; FINC; FN; FN1; FNZ; GFND; GFND2; LETS; MSF
UniProt ID: (Human) P02751
Entrez Gene ID: (Human) 2335
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.