Invitrogen
Manufacturer Code:MA536292
Catalog # PIMA536292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6A15 |
Immunogen | His-dFMR1 fusion protein (Drosophilia melanogaster) (C-terminal 580aa) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AT24755 BcDNA:GM08679 CG6203 CG6203 gene product from transcript CG6203-RC CG6203-PA CG6203-PB CG6203-PC CG6203-PD CG6203-PE CG6203-PF CG6203-PG CG6203-PH CG6203-PI CG6203-PJ CG6203-PK dFMR dfmr1 dFmrp dFXR dFXR1 dFXRP DmelCG6203 Dmel_CG6203 dmfr1 drosophila fragile X mental retardation protein EP(3)3517 FMR FMR1 Fmr-1 Fmr1-PA Fmr1-PB Fmr1-PC Fmr1-PD Fmr1-PE Fmr1-PF Fmr1-PG Fmr1-PH Fmr1-PI Fmr1-PJ Fmr1-PK FMRP fragile X fragile X mental retardation fragile X mental retardation 1 fragile X mental retardation gene fragile X mental retardation protein fragile X mental retardation protein 1 Fragile X mental retardation protein 1 homolog fragile X mental retardation syndrome 1 fragile X mental retardation syndrome 1 homolog Fragile X mental retardation syndrome-related protein 1 fragile X mental retardation-1 protein fragile X protein fragile x related fragile X related protein fragile X-related fragile-X Fragile-X mental retardation protein Fragile-X-related FRAXA FXR MGC87458 POF POF1 protein FMR-1 ragile X mental retardation protein synaptic functional regulator FMR1; cg 6203 gene product from transcript cg6203-rc; CG6203-PA; CG6203-PB; CG6203-PC; CG6203-PD; CG6203-PE; CG6203-PF; CG6203-PG; CG6203-PH; CG6203-PI; CG6203-PJ; CG6203-PK; dFMR; dFMR1; drosophila fragile X mental retardation protein; Fmr1-PA; Fmr1-PB; Fmr1-PC; Fmr1-PD; Fmr1-PE; Fmr1-PF; Fmr1-PG; Fmr1-PH; Fmr1-PI; Fmr1-PJ; Fmr1-PK; fragile X; fragile X mental retardation; fragile X mental retardation 1; fragile X mental retardation gene; fragile X mental retardation protein; fragile X mental retardation protein 1; fragile X mental retardation related 1; Fragile X mental retardation syndrome-related protein 1; fragile X protein; fragile x related; fragile X related protein; fragile X retardation 1 protein; fragile X-related; fragile-X; Fragile-X mental retardation 1; Fragile-X mental retardation protein; Fragile-X-related; FXR; Synaptic functional regulator FMR1
Gene Aliases: AT24755; BcDNA:GM08679; CG6203; dFMR; DFmr1; DFMRP; dFXR; dFXR1; dFXRP; Dmel\CG6203; Dmel_CG6203; dmfr1; EP(3)3517; FMR; FMR1; FMRP; FXR
UniProt ID: (Fruit fly) Q8T0M0
Entrez Gene ID: (Fruit fly) 37528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.