Novus Biologicals
Manufacturer Code:NBP142567
Catalog # NBP142567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G9 |
Immunogen | Purified recombinant fragment of human FMR1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FMRP; FMRPFRAXAPOF fragile X mental retardation 1 fragile X mental retardation 1 protein MGC87458 POF1 premature ovarian failure 1 Protein FMR-1; Fragile X mental retardation protein 1; Synaptic functional regulator FMR1
Gene Aliases: FMR1; FMRP; FRAXA; POF; POF1
UniProt ID: (Human) Q06787
Entrez Gene ID: (Human) 2332
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.