OriGene
Manufacturer Code:TA504291
Catalog # 50-167-5012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 36-279 of human FMR1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FMRP; FMRP FRAXA POF POF1; fragile X mental retardation 1; Fragile X mental retardation protein 1; Fragile X mental retardation protein 1 homolog; fragile X mental retardation syndrome 1 homolog; fragile X mental retardation-1 protein; protein FMR-1; ragile X mental retardation protein; Synaptic functional regulator FMR1
Gene Aliases: Fmr-1; FMR1; FMRP; FRAXA; POF; POF1
UniProt ID: (Human) Q06787, (Rat) Q80WE1
Entrez Gene ID: (Dog) 481070, (Human) 2332, (Rat) 24948, (Mouse) 14265
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.