Abnova
Manufacturer Code:H00002332M01
Catalog # 89-022-198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D4 |
Immunogen | FMR1 (NP_002015 121 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant FMR1.
For Research Use Only
Protein Aliases: FMRP; Fragile X mental retardation protein 1; FRAXA; MGC87458; POF; POF1; Synaptic functional regulator FMR1
Gene Aliases: FMR1; FMRP; FRAXA; POF; POF1
UniProt ID: (Human) Q06787
Entrez Gene ID: (Human) 2332
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.