Invitrogen
Manufacturer Code:PA530035
Catalog # PIPA530035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 202 of Human FMO5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Baeyer-Villiger monooxygenase 1; Dimethylaniline monooxygenase [N-oxide-forming] 5; Dimethylaniline oxidase 5; Flavin-containing monooxygenase 5; FMO 5; FMO 5 dimethylaniline monooxygenase &lsqbN-oxide-forming] 5 dimethylaniline oxidase 5 hepatic flavin-containing monooxygenase 5; hBVMO1; hepatic flavin-containing monooxygenase 5; NAPDH oxidase
Gene Aliases: FMO5
UniProt ID: (Human) P49326
Entrez Gene ID: (Human) 2330
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.