Proteintech
Manufacturer Code:136991AP150UL
Catalog # 50-172-8942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FMO5 Fusion Protein Ag4604 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Baeyer-Villiger monooxygenase 1; Dimethylaniline monooxygenase [N-oxide-forming] 5; Dimethylaniline oxidase 5; Dimethylaniline oxidase 5 FMO 5 FMO5; Flavin-containing monooxygenase 5; FMO 5; hBVMO1; Hepatic flavin-containing monooxygenase 5; NAPDH oxidase
Gene Aliases: 5033418D19Rik; AI195026; FMO5
UniProt ID: (Human) P49326, (Rat) Q8K4C0, (Mouse) P97872
Entrez Gene ID: (Human) 2330, (Rat) 246248, (Mouse) 14263
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.