Invitrogen
Manufacturer Code:PA579276
Catalog # PIPA579276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Flavin containing monooxygenase 4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase &lsqbN-oxide-forming] 4 Dimethylaniline oxidase 4 Hepatic flavin-containing monooxygenase 4 FMO 4 FMO4 FMO2; Dimethylaniline monooxygenase [N-oxide-forming] 4; Dimethylaniline oxidase 4; flavin-containing monooxygenase 4; FMO 4; Hepatic flavin-containing monooxygenase 4
Gene Aliases: D1Ertd532e; FMO2; FMO4
UniProt ID: (Human) P31512, (Mouse) Q8VHG0, (Rat) Q8K4B7
Entrez Gene ID: (Human) 2329, (Mouse) 226564, (Rat) 246247
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.