OriGene
Manufacturer Code:CF810419
Catalog # 50-167-0059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-291 of human FMO3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 3; Dimethylaniline oxidase 3; FMO 3; FMO form 2; FMO II; FMOII TMAU dJ127D3.1; Hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; Trimethylamine monooxygenase
Gene Aliases: AW111792; dJ127D3.1; FMO3; FMOII; TMAU
UniProt ID: (Human) P31513, (Mouse) P97501
Entrez Gene ID: (Human) 2328, (Mouse) 14262
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.