Invitrogen
Manufacturer Code:PA527916
Catalog # PIPA527916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 122 and 394 of Human FMO3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 3; Dimethylaniline oxidase 3; FMO 3; FMO form 2; FMO II; FMOII TMAU dJ127D3.1 FMO 3 FMO II FMO form 2 dimethylaniline monooxygenase &lsqbN-oxide-forming] 3 dimethylaniline oxidase 3 hepatic flavin-containing monooxygenase 3 hepatic flavin-containing monooxygenase-3; Hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; Trimethylamine monooxygenase
Gene Aliases: AW111792; dJ127D3.1; FMO3; FMOII; TMAU
UniProt ID: (Human) P31513, (Mouse) P97501
Entrez Gene ID: (Human) 2328, (Mouse) 14262
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.