Novus Biologicals
Manufacturer Code:NBP186083
Catalog # NBP186083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IYKSVFSNSSKEMMCFPDFPFPDDFPNFMHNSKIQEYIIAFAKEKNLLKYIQFKTFVSSVNKHPDFATTGQWDVTTERDGKKESAVFDAVMVCSGHHVYPNLPK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 3; dimethylaniline monooxygenase [N-oxide-forming] 3 Dimethylaniline oxidase 3 dJ127D3.1 EC 1.14.13.8 flavin containing monooxygenase 3 FMO 3 FMO form 2 FMO II FMOII Hepatic flavin-containing monooxygenase 3 hepatic flavin-containing monooxygenase-3 MGC34400 TMAU; Dimethylaniline oxidase 3; FMO 3; FMO form 2; FMO II; Hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; Trimethylamine monooxygenase
Gene Aliases: dJ127D3.1; FMO3; FMOII; TMAU
UniProt ID: (Human) P31513
Entrez Gene ID: (Human) 2328
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.