Invitrogen
Manufacturer Code:PA5109732
Catalog # PIPA5109732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 210-470 of human FMO2 (NP_001451.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310008D08Rik 2310042I22Rik AW107733 Dimethylaniline monooxygenase [N-oxide-forming] 2 Dimethylaniline oxidase 2 FMO 1B1 FMO 2 FMO1B1 FMO2 Pulmonary flavin-containing monooxygenase 2; Dimethylaniline monooxygenase [N-oxide-forming] 2; Dimethylaniline oxidase 2; flavin containing monooxygenase 2 (non-functional); FMO 1B1; FMO 2; FMO, pulmonary; Pulmonary flavin-containing monooxygenase 2
Gene Aliases: FMO1B1; FMO2
UniProt ID: (Human) Q99518
Entrez Gene ID: (Human) 2327
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.