Novus Biologicals
Manufacturer Code:NBP133318
Catalog # NBP133318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human FMO2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 2; dimethylaniline monooxygenase [N-oxide-forming] 2 Dimethylaniline oxidase 2 EC 1.14.13.8 flavin containing monooxygenase 2 flavin containing monooxygenase 2 (non-functional) FLJ40826 FMO 1B1 FMO 2 FMO pulmonary FMO1B1 Pulmonary flavin-containing monooxygenase 2; Dimethylaniline oxidase 2; flavin containing monooxygenase 2 (non-functional); FMO 1B1; FMO 2; FMO, pulmonary; Pulmonary flavin-containing monooxygenase 2
Gene Aliases: FMO1B1; FMO2
UniProt ID: (Human) Q99518
Entrez Gene ID: (Human) 2327
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.