Proteintech
Manufacturer Code:168611AP150UL
Catalog # 50-173-1225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 2; Dimethylaniline oxidase 2; Dimethylaniline oxidase 2 FMO 1B1 FMO 2 FMO1B1 FMO2; flavin containing monooxygenase 2 (non-functional); FMO 1B1; FMO 2; FMO, pulmonary; Pulmonary flavin-containing monooxygenase 2
Gene Aliases: 2310008D08Rik; 2310042I22Rik; AW107733; FMO1B1; FMO2
UniProt ID: (Human) Q99518, (Mouse) Q8K2I3, (Rat) Q6IRI9
Entrez Gene ID: (Human) 2327, (Mouse) 55990, (Rat) 246245
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.