Invitrogen
Manufacturer Code:PA595285
Catalog # PIPA595285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human FMO1 different from the related mouse sequence by one amino acid and from the related rat sequence by two amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase &lsqbN-oxide-forming] 1 1.14.13.8 Dimethylaniline oxidase 1 Fetal hepatic flavin-containing monooxygenase 1 FMO 1 FMO1; Dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; Hepatic flavin-containing monooxygenase 1
Gene Aliases: Fmo-1; FMO1; RFMO1A
UniProt ID: (Human) Q01740, (Rat) P36365, (Mouse) P50285
Entrez Gene ID: (Human) 2326, (Rat) 25256, (Mouse) 14261
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.