Invitrogen
Manufacturer Code:PA569138
Catalog # PIPA569138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FMO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1; flavin containing monooxygenase 1; Flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; FMO 1 Flavin-containing monooxygenase 1 (fetal liver) dimethylaniline monooxygenase &lsqbN-oxide-forming] 1 dimethylaniline oxidase 1 fetal hepatic flavin-containing monooxygenase 1; FMO 1A1; FMO form 1; FMO1; hepatic flavin-containing monooxygenase (EC 1.14.13.8); hepatic flavin-containing monooxygenase (FMO); Hepatic flavin-containing monooxygenase 1; monooxygenase (FMO) (EC 1.14.13.8)
Gene Aliases: FMO-1; FMO1; RFMO1A
UniProt ID: (Pig) P16549, (Human) Q01740, (Rabbit) P17636, (Dog) Q95LA2, (Rat) P36365, (Mouse) P50285
Entrez Gene ID: (Pig) 397132, (Human) 2326, (Bovine) 508781, (Rabbit) 100009120, (Horse) 100059719, (Dog) 403604, (Rat) 25256, (Mouse) 14261, (Guinea pig) 100723945
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.