Invitrogen
Manufacturer Code:PA521555
Catalog # PIPA521555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 437 of FMO1 (Uniprot IDNo.Q01740) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; FMO 1 Flavin-containing monooxygenase 1 (fetal liver) dimethylaniline monooxygenase &lsqbN-oxide-forming] 1 dimethylaniline oxidase 1 fetal hepatic flavin-containing monooxygenase 1
Gene Aliases: FMO1
UniProt ID: (Human) Q01740
Entrez Gene ID: (Human) 2326
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.