Novus Biologicals
Manufacturer Code:NBP132469
Catalog # NBP132469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human FMO1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 1; dimethylaniline monooxygenase [N-oxide-forming] 1 Dimethylaniline oxidase 1 EC 1.14.13.8 Fetal hepatic flavin-containing monooxygenase 1 flavin containing monooxygenase 1 Flavin-containing monooxygenase 1 (fetal liver) FMO 1; Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1
Gene Aliases: FMO1
UniProt ID: (Human) Q01740
Entrez Gene ID: (Human) 2326
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.