Proteintech
Manufacturer Code:278341AP150UL
Catalog # 50-173-5771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FMNL1 Fusion Protein Ag27312 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C17orf1 C17orf1B CLL associated antigen KW 13 FHOD4 FMNL FMNL1 formin like 1 Formin like protein 1 KW 13 Leukocyte formin; CLL-associated antigen KW-13; formin-like 1; Formin-like protein 1; Leukocyte formin
Gene Aliases: C17orf1; C17orf1B; FHOD4; FMNL; FMNL1; FRL1; KW-13
UniProt ID: (Human) O95466
Entrez Gene ID: (Human) 752
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.