Invitrogen
Manufacturer Code:PA526684
Catalog # PIPA526684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 478-507 amino acids from the C-terminal region of human FLVC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf58 CCT EPV FLVCRL14q MFSD7C PVHH calcium-chelate transporter feline leukemia virus subgroup C receptor-related protein 2; Calcium-chelate transporter; CCT; feline leukemia virus subgroup C cellular receptor family, member 2; Feline leukemia virus subgroup C receptor-related protein 2
Gene Aliases: C14orf58; CCT; EPV; FLVCR2; FLVCRL14q; MFSD7C; PVHH
UniProt ID: (Human) Q9NX90
Entrez Gene ID: (Human) 55640
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.