Novus Biologicals
Manufacturer Code:NB1001481
Catalog # NB1001481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KTVMLSKQSESAI corresponding to C-Terminus according to NP_054772.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ataxia posterior column 1 with retinitis pigmentosa AXPC1 feline leukemia virus subgroup C cellular receptor 1 Feline leukemia virus subgroup C receptor feline leukemia virus subgroup C receptor-related protein 1 FLVCRFLJ33420 hFLVCR MFSD7B PCA PCARP; Feline leukemia virus subgroup C receptor; Feline leukemia virus subgroup C receptor-related protein 1
Gene Aliases: AXPC1; FLVCR; FLVCR1; MFSD7B; PCA; PCARP
UniProt ID: (Human) Q9Y5Y0
Entrez Gene ID: (Human) 28982
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.