Invitrogen
Manufacturer Code:PA579270
Catalog # PIPA579270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse Flt3 / CD135 recombit protein (Position: E62-E295). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD135; Fetal liver kinase 2; FL cytokine receptor; FLK-2; FLT-3; Fms-like tyrosine kinase 3; Receptor-type tyrosine-protein kinase FLT3; Receptor-type tyrosine-protein kinase FLT3 FL cytokine receptor Fetal liver kinase 2 FLK-2 Fms-like tyrosine kinase 3 FLT-3 Tyrosine-protein kinase receptor flk-2 CD135 Flt3 Flk-2 Flt-3; tyrosine-protein kinase FLT3; Tyrosine-protein kinase receptor flk-2
Gene Aliases: B230315G04; CD135; Flk-2; Flk2; Flt-3; Flt3; Ly72; wmfl
UniProt ID: (Mouse) Q00342
Entrez Gene ID: (Rat) 140635, (Mouse) 14255
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.