Abnova
Manufacturer Code:PAB18232
Catalog # 89-119-802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human FLT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of FLT1.
For Research Use Only
Protein Aliases: FLT; FLT-1; Fms-like tyrosine kinase 1; fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; Vascular permeability factor receptor; VEGFR-1; VEGFR1
Gene Aliases: FLT; FLT-1; FLT1; FRT; VEGFR-1; VEGFR1
UniProt ID: (Human) P17948
Entrez Gene ID: (Human) 2321
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.