Invitrogen
Manufacturer Code:MA530440
Catalog # PIMA530440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11B5G9 |
Immunogen | Recombit Human FLRT2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Fibronectin-like domain-containing leucine-rich transmembrane protein 2; fibronectin-like domain-containing leucine-rich transmembrane protein 2 leucine-rich repeat transmembrane protein FLRT2; Leucine-rich repeat transmembrane protein FLRT2
Gene Aliases: FLRT2; KIAA0405; UNQ232/PRO265
UniProt ID: (Human) O43155
Entrez Gene ID: (Human) 23768
Molecular Function: cytokine extracellular matrix protein receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.