Invitrogen
Manufacturer Code:PA532122
Catalog # PIPA532122
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 58 and 256 of Human FLRT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fibronectin-like domain-containing leucine-rich transmembrane protein 2; fibronectin-like domain-containing leucine-rich transmembrane protein 2 leucine-rich repeat transmembrane protein FLRT2; Leucine-rich repeat transmembrane protein FLRT2
Gene Aliases: FLRT2; KIAA0405; UNQ232/PRO265
UniProt ID: (Human) O43155
Entrez Gene ID: (Human) 23768
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.