Novus Biologicals
Manufacturer Code:NBP298377AF594
Catalog # NB082290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant FLRT2 (Q8BLU0 Met1-Ser539). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fibronectin Leucine Rich Transmembrane Protein 2 Fibronectin-Like Domain-Containing Leucine-Rich Transmembrane Protein 2 KIAA0405 Leucine-Rich Repeat Transmembrane Protein FLRT2; Fibronectin-like domain-containing leucine-rich transmembrane protein 2; Leucine-rich repeat transmembrane protein FLRT2
Gene Aliases: FLRT2; KIAA0405; UNQ232/PRO265
UniProt ID: (Human) O43155
Entrez Gene ID: (Human) 23768
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.