R&D Systems
Manufacturer Code:AF2877SP
Catalog # AF2877SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human FLRT2 Cys36-Ser539 Accession # O43155 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Fibronectin Leucine Rich Transmembrane Protein 2 Fibronectin-Like Domain-Containing Leucine-Rich Transmembrane Protein 2 KIAA0405 Leucine-Rich Repeat Transmembrane Protein FLRT2; Fibronectin-like domain-containing leucine-rich transmembrane protein 2; Leucine-rich repeat transmembrane protein FLRT2
Gene Aliases: FLRT2; KIAA0405; UNQ232/PRO265
UniProt ID: (Human) O43155
Entrez Gene ID: (Human) 23768
Molecular Function: cytokine extracellular matrix protein receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.