Invitrogen
Manufacturer Code:PA547106
Catalog # PIPA547106
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Follistatin-related Gene Protein/FLRG Val24-Val256 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Follistatin-like 3 FSTL3 FLRGFSRP follistatin-like 3 (secreted glycoprotein) Follistatin-like protein 3 Follistatin-related gene protein follistatin-related protein 3; Follistatin-like protein 3; Follistatin-related gene protein; Follistatin-related protein 3
Gene Aliases: E030038F23Rik; Flrg; Fstl3
UniProt ID: (Mouse) Q9EQC7
Entrez Gene ID: (Mouse) 83554
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.