Abnova
Manufacturer Code:H00002319M03
Catalog # 89-022-188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G6 |
Immunogen | FLOT2 (AAH17292 1 a.a. ~ 379 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FLOT2.
For Research Use Only
Protein Aliases: ECS-1; ECS1; Epidermal surface antigen; ESA; ESA1; Flotillin 2 (epidermal surface antigen 1); Flotillin-2; M17S1; Membrane component chromosome 17 surface marker 1; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1)
Gene Aliases: ECS-1; ECS1; ESA; ESA1; FLOT2; M17S1
UniProt ID: (Human) Q14254
Entrez Gene ID: (Human) 2319
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.