Invitrogen
Manufacturer Code:MA531679
Catalog # PIMA531679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7A3D12 |
Immunogen | Purified recombit fragment of human CFLAR (AA: 100-251) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-FLIP; CASH; CASH FLIP MRIT CLARP FLAME Casper FLAME1 c-FLIP FLAME-1 I-FLICE c-FLIPL c-FLIPR c-FLIPS CASP8AP1; CASP8 and FADD-like apoptosis regulator; CASP8 and FADD-like apoptosis regulator subunit p12; CASP8 and FADD-like apoptosis regulator subunit p43; Caspase homolog; Caspase-eight-related protein; Caspase-like apoptosis regulatory protein; caspase-related inducer of apoptosis; Casper; Cellular FLICE-like inhibitory protein; CLARP; FADD-like antiapoptotic molecule 1; FLAME-1; I-FLICE; Inhibitor of FLICE; MACH-related inducer of toxicity; MRIT; testis secretory sperm-binding protein Li 233m; Usurpin; usurpin beta
Gene Aliases: 2310024N18Rik; A430105C05Rik; c-FLIP; c-FLIPL; c-FLIPR; c-FLIPS; CASH; CASP8AP1; Casper; CFLAR; CLARP; ENSMUSG00000072980; FLAME; FLAME-1; FLAME1; FLIP; Gm9845; I-FLICE; MRIT
UniProt ID: (Human) O15519, (Mouse) O35732
Entrez Gene ID: (Human) 8837, (Mouse) 12633
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.