Novus Biologicals
Manufacturer Code:NB10056306
Catalog # NB10056306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to amino acids 206-222 GRSKEQRLKEQLGAQQE of human I-FLICE was used to immunize rabbits. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-FLIP; CASH; CASHFADD-like antiapoptotic molecule 1 CASP8 and FADD-like apoptosis regulator CASP8AP1 Caspase homolog Caspase-eight-related protein Caspase-like apoptosis regulatory protein caspase-related inducer of apoptosis CASPER CasperFLAME1 Cellular FLICE-like inhibitory protein c-FLIPR c-FLIPS CLARPMACH-related inducer of toxicity FADD-like apoptotic molecule FLAME FLAME-1 I-FLICEMRITc-FLIPFLIPc-FLIPL Inhibitor of FLICE Usurpin usurpin beta; CASP8 and FADD-like apoptosis regulator; CASP8 and FADD-like apoptosis regulator subunit p12; CASP8 and FADD-like apoptosis regulator subunit p43; Caspase homolog; Caspase-eight-related protein; Caspase-like apoptosis regulatory protein; caspase-related inducer of apoptosis; Casper; Cellular FLICE-like inhibitory protein; CLARP; FADD-like antiapoptotic molecule 1; FLAME-1; I-FLICE; Inhibitor of FLICE; MACH-related inducer of toxicity; MRIT; testis secretory sperm-binding protein Li 233m; Usurpin; usurpin beta
Gene Aliases: c-FLIP; c-FLIPL; c-FLIPR; c-FLIPS; CASH; CASP8AP1; Casper; CFLAR; CLARP; FLAME; FLAME-1; FLAME1; FLIP; I-FLICE; MRIT
UniProt ID: (Human) O15519
Entrez Gene ID: (Human) 8837
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.