Novus Biologicals
Manufacturer Code:NB500219
Catalog # NB500219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C-terminus of the FLASH protein (proprietary peptide). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CASP8 associated protein 2; CASP8 associated protein 2 caspase 8 associated protein 2 CED-4 FLASHKIAA1315CASP8-associated protein 2 FLICE associated huge FLICE-associated huge protein FLJ11208 human FLASH RIP25FLASH homolog RIP25; CASP8-associated protein 2; CASP8AP2; FLASH homolog RIP25; FLICE associated huge; FLICE-associated huge protein; human FLASH
Gene Aliases: CASP8AP2; CED-4; FLASH; KIAA1315; RIP25
UniProt ID: (Human) Q9UKL3
Entrez Gene ID: (Human) 9994
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.